Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004200.1 | NP_001004200.1 | 9 - 10 | 1073 | 114 | |
GenBank mRNA | BC079143.1 | - | 9 - 10 | 1073 | 7128107 | 114 |
FQ213575.1 | - | 5 - 6 | 607 | 114 | ||
FQ213666.1 | - | 5 - 6 | 604 | 114 | ||
FQ228882.1 | - | 9 - 10 | 1114 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |