Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768223.1 | XP_008766445.1 | 6 - 7 | 144 | ||
XM_008768224.1 | XP_008766446.1 | 7 - 8 | 144 | |||
XM_008768225.1 | XP_008766447.1 | 6 - 7 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |