Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053732.1 | NP_446184.1 | 5 - 6 | 1937 | 100 | |
XM_008767531.1 | XP_008765753.1 | 5 - 6 | 3518 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |