Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109031.1 | NP_001102501.1 | 1 - 2 | 114 | 106 | |
XM_006246394.2 | XP_006246456.1 | 1 - 2 | 335 | 106 | ||
XM_006246395.2 | XP_006246457.1 | 1 - 2 | 333 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |