Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107053.1 | NP_001100523.1 | 3 - 4 | 972 | 85 | |
XM_006247328.2 | XP_006247390.1 | 4 - 5 | 1792 | 85 | ||
XM_008768055.1 | XP_008766277.1 | 4 - 5 | 1792 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |