Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001062937.5 | XP_001062937.3 | 7 - 8 | 851 | 57 | |
XM_006247478.2 | XP_006247540.1 | 5 - 6 | 1559 | 57 | ||
XM_006247479.2 | XP_006247541.1 | 8 - 9 | 751 | 57 | ||
XM_006247480.2 | XP_006247542.2 | 3 - 4 | 273 | 57 | ||
XM_008768226.1 | XP_008766448.1 | 5 - 6 | 1827 | 57 | ||
XM_008768227.1 | XP_008766449.1 | 7 - 8 | 2781 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |