Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031833.2 | NP_114021.2 | 1 - 2 | 164 | 102 | |
GenBank mRNA | BC086599.1 | - | 1 - 2 | 164 | 7312852 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |