Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107286.1 | NP_001100756.1 | 4 - 5 | 396 | 162 | |
GenBank mRNA | BC166991.1 | - | 1 - 2 | 66 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |