Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252478.2 | XP_006252540.1 | 11 - 12 | 1187 | 63 | |
XM_224534.8 | XP_224534.7 | 12 - 13 | 1433 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Probe spans exons |