Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148891.2 | NP_683689.1 | 3 - 4 | 769 | 105 | |
GenBank mRNA | AJ492222.1 | - | 3 - 4 | 390 | 105 | |
BC097277.1 | - | 3 - 4 | 769 | 7457194 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |