Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044224.1 | NP_001037689.1 | 19 - 20 | 3152 | 65 | |
GenBank mRNA | AB231661.1 | - | 19 - 20 | 3152 | 65 | |
AF111111.1 | - | 7 - 8 | 724 | 65 | ||
BC166408.1 | - | 19 - 20 | 3142 | 9030944 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |