Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173128.1 | NP_775151.1 | 2 | 500 | 57 | |
GenBank mRNA | M63665.1 | - | 2 | 500 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |