Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ublcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014117.1 | NP_001014139.1 | 9 - 10 | 879 | 153 | |
XM_006246140.1 | XP_006246202.1 | 9 - 10 | 909 | 153 | ||
GenBank mRNA | BC089210.1 | - | 9 - 10 | 879 | 7302690 | 153 |
FQ233702.1 | - | 9 - 10 | 882 | 153 | ||
FQ233841.1 | - | 9 - 10 | 882 | 153 | ||
FQ235222.1 | - | 1 - 2 | 97 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |