Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247940.2 | XP_006248002.1 | 10 - 11 | 1216 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |