Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139115.2 | NP_620815.2 | 6 - 7 | 886 | 120 | |
XM_006246885.2 | XP_006246947.1 | 7 - 8 | 1963 | 120 | ||
XM_006246886.2 | XP_006246948.1 | 7 - 8 | 1965 | 120 | ||
XM_006246887.2 | XP_006246949.1 | 7 - 8 | 1963 | 120 | ||
XM_006246888.2 | XP_006246950.1 | 7 - 8 | 1965 | 120 | ||
XM_006246889.2 | XP_006246951.1 | 6 - 7 | 1843 | 120 | ||
XM_006246890.2 | XP_006246952.1 | 6 - 7 | 1843 | 120 | ||
XM_006246891.2 | XP_006246953.1 | 7 - 8 | 1964 | 120 | ||
GenBank mRNA | AF140359.1 | - | 7 - 8 | 1259 | 120 | |
BC080241.1 | - | 6 - 7 | 2425 | 120 | ||
BC098656.1 | - | 6 - 7 | 886 | 7192854 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |