Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107048.1 | NP_001100518.1 | 2 - 3 | 1033 | 143 | |
XM_006247304.2 | XP_006247366.1 | 2 - 3 | 1352 | 143 | ||
GenBank mRNA | FQ228290.1 | - | 2 - 3 | 987 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |