Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019156.2 | NP_062029.2 | 7 - 8 | 1407 | 73 | |
GenBank mRNA | AY318963.1 | - | 1404 | 73 | ||
BC105821.1 | - | 7 - 8 | 1407 | 7371104 | 73 | |
FQ209609.1 | - | 7 - 8 | 1420 | 73 | ||
FQ209682.1 | - | 7 - 8 | 1415 | 73 | ||
FQ209762.1 | - | 3 - 4 | 581 | 73 | ||
FQ212575.1 | - | 7 - 8 | 1407 | 73 | ||
FQ218401.1 | - | 5 - 6 | 979 | 73 | ||
FQ219297.1 | - | 7 - 8 | 1421 | 73 | ||
U44845.1 | - | 7 - 8 | 1397 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |