Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013975.2 | NP_001013997.2 | 2 - 3 | 438 | 89 | |
XM_006247887.2 | XP_006247949.1 | 2 - 3 | 671 | 89 | ||
GenBank mRNA | DV215880.1 | - | 2 - 3 | 438 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |