Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246472.2 | XP_006246534.1 | 2 - 3 | 235 | 81 | |
XM_008767786.1 | XP_008766008.1 | 1 - 2 | 223 | 81 | ||
GenBank mRNA | EF445633.1 | - | 2 - 3 | 139 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |