Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053508.2 | NP_445960.1 | 2 - 3 | 271 | 82 | |
GenBank mRNA | AJ306427.1 | - | 1 - 2 | 200 | 82 | |
BC078773.1 | - | 2 - 3 | 271 | 7133616 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |