Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105815.1 | NP_001099285.1 | 4 - 5 | 626 | 95 | |
XM_006246935.2 | XP_006246997.1 | 3 - 4 | 440 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |