Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108276.1 | NP_001101746.1 | 1 - 2 | 541 | 147 | |
XM_006246505.2 | XP_006246567.1 | 1 - 2 | 541 | 147 | ||
GenBank mRNA | BC160894.1 | - | 1 - 2 | 492 | 7446534 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |