Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108276.1 | NP_001101746.1 | 4 - 5 | 890 | 70 | |
XM_006246505.2 | XP_006246567.1 | 4 - 5 | 889 | 70 | ||
GenBank mRNA | BC160894.1 | - | 4 - 5 | 840 | 7446534 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |