Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008818.1 | NP_001008818.1 | 4 - 5 | 882 | 144 | |
XM_006247394.2 | XP_006247456.1 | 4 - 5 | 906 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |