Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246843.2 | XP_006246905.1 | 2 - 3 | 253 | 106 | |
XM_006246844.2 | XP_006246906.1 | 1 - 2 | 77 | 106 | ||
GenBank mRNA | CB605808.1 | - | 1 - 2 | 54 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |