Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007622.1 | NP_001007623.1 | 3 - 4 | 364 | 55 | |
XM_006252255.2 | XP_006252317.1 | 3 - 4 | 382 | 55 | ||
XM_006252256.2 | XP_006252318.1 | 3 - 4 | 374 | 55 | ||
XM_006252257.2 | XP_006252319.1 | 3 - 4 | 896 | 55 | ||
XM_006252258.1 | XP_006252320.1 | 3 - 4 | 264 | 55 | ||
XM_006252259.2 | XP_006252321.1 | 3 - 4 | 931 | 55 | ||
XM_008770823.1 | XP_008769045.1 | 3 - 4 | 265 | 55 | ||
GenBank mRNA | AY531526.1 | - | 3 - 4 | 346 | 55 | |
BC079091.1 | - | 3 - 4 | 364 | 7123600 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |