Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106050.1 | NP_001099520.1 | 2 - 3 | 313 | 85 | |
GenBank mRNA | FQ229062.1 | - | 2 - 3 | 187 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |