Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034937.2 | NP_001030109.2 | 1 - 2 | 160 | 77 | |
XM_006252509.2 | XP_006252571.1 | 1 - 2 | 234 | 77 | ||
GenBank mRNA | CB708329.1 | - | 1 - 2 | 171 | 77 | |
FM053767.1 | - | 1 - 2 | 88 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |