Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013051.1 | NP_001013069.1 | 6 - 7 | 427 | 115 | |
GenBank mRNA | BC158724.1 | - | 6 - 7 | 427 | 9090831 | 115 |
FQ220550.1 | - | 6 - 7 | 462 | 115 | ||
FQ221379.1 | - | 6 - 7 | 453 | 115 | ||
FQ225182.1 | - | 6 - 7 | 545 | 115 | ||
FQ227975.1 | - | 6 - 7 | 545 | 115 | ||
FQ232174.1 | - | 6 - 7 | 545 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |