Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013051.1 | NP_001013069.1 | 10 - 11 | 686 | 138 | |
GenBank mRNA | BC158724.1 | - | 10 - 11 | 686 | 9090831 | 138 |
FQ220550.1 | - | 10 - 11 | 721 | 138 | ||
FQ225182.1 | - | 10 - 11 | 804 | 138 | ||
FQ227975.1 | - | 10 - 11 | 804 | 138 | ||
FQ232174.1 | - | 10 - 11 | 804 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |