Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053520.2 | NP_445972.2 | 6 - 7 | 1029 | 169 | |
XM_006252347.2 | XP_006252409.1 | 7 - 8 | 2312 | 169 | ||
XM_006252348.2 | XP_006252410.1 | 6 - 7 | 1050 | 169 | ||
XM_006252349.2 | XP_006252411.1 | 8 - 9 | 2478 | 169 | ||
XM_006252351.2 | XP_006252413.1 | 7 - 8 | 1556 | 169 | ||
XM_006252352.2 | XP_006252414.1 | 7 - 8 | 1516 | 169 | ||
XM_006252354.2 | XP_006252416.1 | 6 - 7 | 2204 | 169 | ||
XM_008770898.1 | XP_008769120.1 | 7 - 8 | 3816 | 169 | ||
XM_008770899.1 | XP_008769121.1 | 7 - 8 | 1512 | 169 | ||
XM_008770900.1 | XP_008769122.1 | 7 - 8 | 1068 | 169 | ||
XM_008770901.1 | XP_008769123.1 | 7 - 8 | 3792 | 169 | ||
XM_008770902.1 | XP_008769124.1 | 6 - 7 | 3708 | 169 | ||
XM_008770903.1 | XP_008769125.1 | 6 - 7 | 3684 | 169 | ||
GenBank mRNA | AB030216.1 | - | 5 - 6 | 690 | 169 | |
AB030217.1 | - | 4 - 5 | 525 | 169 | ||
AB046531.1 | - | 6 - 7 | 808 | 169 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |