Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053520.2 | NP_445972.2 | 3 - 4 | 588 | 77 | |
XM_006252347.2 | XP_006252409.1 | 4 - 5 | 1847 | 77 | ||
XM_006252348.2 | XP_006252410.1 | 3 - 4 | 585 | 77 | ||
XM_006252349.2 | XP_006252411.1 | 5 - 6 | 2013 | 77 | ||
XM_006252351.2 | XP_006252413.1 | 4 - 5 | 1091 | 77 | ||
XM_006252352.2 | XP_006252414.1 | 4 - 5 | 1051 | 77 | ||
XM_008770898.1 | XP_008769120.1 | 4 - 5 | 3351 | 77 | ||
XM_008770899.1 | XP_008769121.1 | 4 - 5 | 1047 | 77 | ||
XM_008770900.1 | XP_008769122.1 | 4 - 5 | 603 | 77 | ||
XM_008770901.1 | XP_008769123.1 | 4 - 5 | 3351 | 77 | ||
GenBank mRNA | AB046531.1 | - | 3 - 4 | 367 | 77 | |
FQ221456.1 | - | 5 - 6 | 1156 | 77 | ||
FQ233920.1 | - | 4 - 5 | 662 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |