Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012773.2 | NP_036905.2 | 3 - 4 | 233 | 112 | |
XM_006252344.1 | XP_006252406.1 | 4 - 5 | 407 | 112 | ||
XM_008770886.1 | XP_008769108.1 | 4 - 5 | 1299 | 112 | ||
XM_008770887.1 | XP_008769109.1 | 4 - 5 | 489 | 112 | ||
XM_008770888.1 | XP_008769110.1 | 4 - 5 | 489 | 112 | ||
XM_008770889.1 | XP_008769111.1 | 4 - 5 | 489 | 112 | ||
XM_008770890.1 | XP_008769112.1 | 4 - 5 | 489 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |