Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108384.1 | NP_001101854.1 | 3 - 4 | 419 | 69 | |
XM_006252275.2 | XP_006252337.1 | 3 - 4 | 461 | 69 | ||
XM_006252276.2 | XP_006252338.1 | 3 - 4 | 864 | 69 | ||
XM_006252277.2 | XP_006252339.1 | 3 - 4 | 461 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |