Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199269.1 | NP_954890.1 | 7 - 8 | 1498 | 113 | |
XM_008764688.1 | XP_008762910.1 | 6 - 7 | 2287 | 113 | ||
GenBank mRNA | AY371924.1 | - | 7 - 8 | 1498 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |