Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114180.1 | NP_001107652.1 | 6 - 7 | 791 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |