Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185025.1 | NP_001171954.1 | 2 - 3 | 268 | 79 | |
XM_008764655.1 | XP_008762877.1 | 2 - 3 | 223 | 79 | ||
GenBank mRNA | BP486352.1 | - | 2 - 3 | 274 | 79 | |
CO399813.1 | - | 2 - 3 | 244 | 79 | ||
FQ210674.1 | - | 2 - 3 | 286 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |