Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106736.2 | NP_001100206.2 | 4 - 5 | 472 | 103 | |
GenBank mRNA | BC168873.1 | - | 4 - 5 | 437 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |