Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cript | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019907.1 | NP_063972.1 | 4 - 5 | 307 | 150 | |
GenBank mRNA | AF047384.1 | - | 4 - 5 | 307 | 150 | |
FQ212096.1 | - | 1 - 2 | 52 | 150 | ||
FQ222669.1 | - | 4 - 5 | 340 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |