Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nova1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100541.1 | NP_001094011.1 | 2 - 3 | 275 | 74 | |
XM_006240069.2 | XP_006240131.1 | 2 - 3 | 729 | 74 | ||
XM_006240070.2 | XP_006240132.1 | 2 - 3 | 613 | 74 | ||
GenBank mRNA | AY262017.1 | - | 2 - 3 | 275 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |