Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007613.1 | NP_001007614.1 | 12 - 13 | 1446 | 137 | |
XM_008768312.1 | XP_008766534.1 | 11 - 12 | 1435 | 137 | ||
XM_008768313.1 | XP_008766535.1 | 11 - 12 | 1358 | 137 | ||
GenBank mRNA | BC079036.1 | - | 12 - 13 | 1446 | 7114313 | 137 |
FJ168767.1 | - | 11 - 12 | 1277 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |