Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42se2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126089.2 | NP_001119561.1 | 1 - 2 | 84 | 120 | |
GenBank mRNA | BC129107.1 | - | 1 - 2 | 84 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |