Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108307.1 | NP_001101777.1 | 1 - 2 | 100 | 120 | |
GenBank mRNA | BC168942.1 | - | 1 - 2 | 87 | 8365819 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |