Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107022.1 | NP_001100492.1 | 803 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |