Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768149.1 | XP_008766371.1 | 18 - 19 | 1761 | 99 | |
GenBank mRNA | FQ210246.1 | - | 18 - 19 | 1729 | 99 | |
FQ217168.1 | - | 18 - 19 | 1725 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |