Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002022.1 | NP_001002022.1 | 5 - 6 | 612 | 107 | |
XM_006246149.1 | XP_006246211.1 | 5 - 6 | 606 | 107 | ||
XM_008767688.1 | XP_008765910.1 | 4 - 5 | 5102 | 107 | ||
XM_008767689.1 | XP_008765911.1 | 5 - 6 | 514 | 107 | ||
GenBank mRNA | BC076397.1 | - | 5 - 6 | 612 | 7109965 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |