Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247961.2 | XP_006248023.1 | 3 - 4 | 516 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |