Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031603.1 | NP_113791.1 | 1 - 2 | 152 | 113 | |
GenBank mRNA | BC063163.1 | - | 1 - 2 | 130 | 6921685 | 113 |
FQ220393.1 | - | 1 - 2 | 158 | 113 | ||
M84416.1 | - | 1 - 2 | 132 | 113 | ||
U53882.1 | - | 1 - 2 | 152 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |