Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013969.1 | NP_001013991.1 | 1 - 2 | 265 | 73 | |
XM_006246939.2 | XP_006247001.1 | 2 - 3 | 573 | 73 | ||
GenBank mRNA | BC089868.1 | - | 1 - 2 | 265 | 7322786 | 73 |
FQ231269.1 | - | 1 - 2 | 291 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |