Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108287.1 | NP_001101757.1 | 5 - 6 | 721 | 153 | |
XM_008768073.1 | XP_008766295.1 | 5 - 6 | 878 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |